Assay Details
| Gene Symbol: B4GALNT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178537.4 | NP_848632.2 | 8 - 9 | 794 | 84 | |
| XM_017017654.1 | XP_016873143.1 | 8 - 9 | 2106 | 84 | ||
| GenBank mRNA | AB089939.1 | - | 8 - 9 | 785 | 84 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000015.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000530717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000530717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000530717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000530717.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000202.hg.1 | ENST00000329962.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000202.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11000202.hg.1 | TC11000015.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1100144376.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100144376.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100144376.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100144376.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100144376.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100144376.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100144376.hg.1 | TC1100006455.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000329962.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000524443.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000526584.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000530717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000530717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000530717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000530717.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | ENST00000534778.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006455.hg.1 | NM_178537.4 | Refseq |

B4GALNT4
Genomic Map