Assay Details
| Gene Symbol: FBXL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001321213.1 | NP_001308142.1 | 4 - 5 | 2127 | 79 | |
| GenBank mRNA | AK021529.1 | - | 2 - 3 | 734 | 79 | |
| AK095394.1 | - | 2 - 3 | 1654 | 79 | ||
| AK294949.1 | - | 4 - 5 | 2121 | 79 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007490.hg.1 | ENST00000578650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007490.hg.1 | ENST00000578650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007490.hg.1 | ENST00000578650.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700007490.hg.1 | ENST00000578650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007490.hg.1 | ENST00000578650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007490.hg.1 | ENST00000578650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007490.hg.1 | ENST00000578650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700007490.hg.1 | ENST00000578650.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000382368.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000415009.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000453700.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000458142.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | ENST00000620087.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700010162.hg.1 | NM_024963.5 | Refseq |

FBXL18
Genomic Map