Assay Details
| Gene Symbol: CLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286105.1 | NP_001273034.1 | 3 - 4 | 542 | 65 | |
| GenBank mRNA | AF015598.1 | - | 4 - 5 | 403 | 65 | |
| AF015600.1 | - | 3 - 4 | 325 | 65 | ||
| AF015602.1 | - | 2 - 3 | 189 | 65 | ||
| AF077967.1 | - | 4 - 5 | 298 | 65 | ||
| AF077973.1 | - | 3 - 4 | 219 | 65 | ||
| AK294250.1 | - | 5 - 6 | 427 | 65 | ||
| AK297690.1 | - | 3 - 4 | 484 | 65 | ||
More Information
Set Membership: |
|

CLN3
Genomic Map