Assay Details
| Gene Symbol: CLN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF015596.1 | - | 2 - 3 | 192 | 60 | |
| AF015599.1 | - | 3 - 4 | 287 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF015596.1 transcripts.
Set Membership: |
|

CLN3
Genomic Map