| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022816.hg.1 |
TC08001732.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022816.hg.1 |
ENST00000320476.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022816.hg.1 |
ENST00000356994.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022816.hg.1 |
ENST00000377533.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022816.hg.1 |
NM_015356.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR08022816.hg.1 |
NM_182706.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000320476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000356994.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000377533.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000525051.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000526832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000531163.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
NM_015356.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
NM_182706.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000320476.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000356994.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000377533.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000525051.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000526832.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
ENST00000531163.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
NM_015356.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
NM_182706.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0800183007.hg.1 |
TC0800012165.hg.1 |
Clariom D TC |