Assay Details
| Gene Symbol: SF3B3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012426.4 | NP_036558.3 | 12 - 13 | 1765 | 72 | |
| GenBank mRNA | AJ001443.1 | - | 12 - 13 | 1791 | 72 | |
| AK074933.1 | - | 6 - 7 | 660 | 72 | ||
| AK291768.1 | - | 12 - 13 | 1763 | 72 | ||
| BC068974.1 | - | 12 - 13 | 1702 | 6168677 | 72 | |
| D87686.1 | - | 12 - 13 | 1710 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC16000579.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000302516.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000562722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000562722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000562722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000562722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000562722.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000562874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000562874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000562874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000562874.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000563739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000563739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000563739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000563739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000563739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000563739.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000564899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000564899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000564899.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000565990.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000565990.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000565990.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000565990.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000565990.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000565990.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000566095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000566095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000566095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000566095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000566095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000566095.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567250.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567635.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000567654.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000568539.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000569687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000569687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000569687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000572365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000572365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000572365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000572365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000572365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000572365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000572365.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000577085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000577085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000577085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000577085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000577085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000577085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | ENST00000577085.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600008275.hg.1 | NM_012426.4 | Refseq |

SF3B3
Genomic Map