Assay Details
Gene Symbol: SF3B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012426.4 | NP_036558.3 | 23 - 24 | 3476 | 66 | |
GenBank mRNA | AJ001443.1 | - | 23 - 24 | 3502 | 66 | |
AK001297.1 | - | 12 - 13 | 1855 | 66 | ||
AK074933.1 | - | 17 - 18 | 2371 | 66 | ||
AK291768.1 | - | 23 - 24 | 3474 | 66 | ||
AL110251.1 | - | 2 - 3 | 260 | 66 | ||
BC000463.1 | - | 5 - 6 | 820 | 66 | ||
BC003146.1 | - | 5 - 6 | 825 | 66 | ||
BC068974.1 | - | 23 - 24 | 3413 | 6168677 | 66 | |
D13642.1 | - | 6 - 7 | 947 | 66 | ||
D87686.1 | - | 23 - 24 | 3421 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16000579.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
