Assay Details
| Gene Symbol: SNX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012084.1 | XP_016867573.1 | 1 - 2 | 263 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017012084.1 transcripts.
Set Membership: |
|

SNX8
Genomic Map