Assay Details
| Gene Symbol: PACSIN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199583.2 | NP_001186512.1 | 1 - 2 | 199 | 141 | |
| GenBank mRNA | AK294132.1 | - | 1 - 2 | 199 | 141 | |
| BC040228.1 | - | 1 - 2 | 184 | 5173129 | 141 | |
| BP251080.1 | - | 1 - 2 | 199 | 141 | ||
| DC312410.1 | - | 1 - 2 | 199 | 141 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000504.hg
Additional Information:
This assay is in the 5'UTR region of the AK294132.1,BC040228.1,NM_001199583.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
regulation of endocytosis
negative regulation of endocytosis
synaptic vesicle endocytosis
neuron projection morphogenesis
protein localization to membrane
establishment of protein localization to plasma membrane
membrane tubulation
positive regulation of dendrite development
protein binding
phospholipid binding
cytoskeletal protein binding
actin filament organization
regulation of endocytosis
negative regulation of endocytosis
synaptic vesicle endocytosis
neuron projection morphogenesis
protein localization to membrane
establishment of protein localization to plasma membrane
membrane tubulation
positive regulation of dendrite development
protein binding
phospholipid binding
cytoskeletal protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005274.hg.1 | TC06000504.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005274.hg.1 | ENST00000436831.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005274.hg.1 | ENST00000486120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005274.hg.1 | ENST00000538621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06005274.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153769.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153769.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153769.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153769.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153769.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153769.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153769.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600153769.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600153769.hg.1 | TC0600007756.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000244458.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000374043.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000486120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000487760.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000493633.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000538621.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | ENST00000620693.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_001199583.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600007756.hg.1 | NM_020804.4 | Refseq |

PACSIN1
Genomic Map