Assay Details
| Gene Symbol: C6orf25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138274.2 | NP_612118.1 | 2 - 3 | 453 | 138 | |
| GenBank mRNA | AJ292262.1 | - | 2 - 3 | 413 | 138 | |
More Information
Set Membership: |
|

C6orf25
Genomic Map