Assay Details
Gene Symbol: IZUMO4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005259480.3 | XP_005259537.2 | 7 - 8 | 113 | ||
GenBank mRNA | AW182303.1 | - | 1 - 2 | 332 | 113 |

More Information
Set Membership: |
![]() ![]() ![]() |
