Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012151.hg.1 |
TC07000764.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012151.hg.1 |
ENST00000354725.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012151.hg.1 |
ENST00000438400.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR07012151.hg.1 |
NM_014390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164014.hg.1 |
ENST00000354725.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164014.hg.1 |
ENST00000489417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164014.hg.1 |
NM_014390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164014.hg.1 |
ENST00000354725.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164014.hg.1 |
ENST00000489417.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0700164014.hg.1 |
NM_014390.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0700164014.hg.1 |
TC0700009020.hg.1 |
Clariom D TC |