Assay Details
| Gene Symbol: RPS3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005.4 | NP_000996.2 | 2 - 3 | 215 | 70 | |
| NM_001256802.1 | NP_001243731.1 | 2 - 3 | 215 | 70 | ||
| NM_001260506.1 | NP_001247435.1 | 2 - 3 | 215 | 70 | ||
| GenBank mRNA | AB062288.1 | - | 2 - 3 | 151 | 70 | |
| AK129794.1 | - | 2 - 3 | 187 | 70 | ||
| AK130960.1 | - | 2 - 3 | 188 | 70 | ||
| AK223048.1 | - | 2 - 3 | 164 | 70 | ||
| AK313051.1 | - | 2 - 3 | 184 | 70 | ||
| BC003137.1 | - | 2 - 3 | 164 | 2906158 | 70 | |
| BC003577.1 | - | 2 - 3 | 156 | 70 | ||
| BC013196.1 | - | 2 - 3 | 156 | 70 | ||
| BC013231.1 | - | 2 - 3 | 176 | 70 | ||
| BC029981.1 | - | 1 - 2 | 109 | 70 | ||
| BC034149.1 | - | 2 - 3 | 188 | 4665438 | 70 | |
| BC071917.1 | - | 2 - 3 | 167 | 6297012 | 70 | |
| BC100284.1 | - | 2 - 3 | 176 | 6160514 | 70 | |
| BQ645330.1 | - | 175 | 70 | |||
| S42658.1 | - | 2 - 3 | 166 | 70 | ||
| U14990.1 | - | 2 - 3 | 158 | 70 | ||
| U14991.1 | - | 2 - 3 | 158 | 70 | ||
| U14992.1 | - | 2 - 3 | 158 | 70 | ||
| X55715.1 | - | 2 - 3 | 180 | 70 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC017103.1 | - | 980 | Chr. - - [-] on Build | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB062288.1 transcripts.
Set Membership: |
|

RPS3
Genomic Map