Assay Details
| Gene Symbol: KIF5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004521.2 | NP_004512.1 | 11 - 12 | 1582 | 100 | |
| XM_017016224.1 | XP_016871713.1 | 11 - 12 | 1188 | 100 | ||
| GenBank mRNA | AK289413.1 | - | 11 - 12 | 1528 | 100 | |
| AK299542.1 | - | 2311 | 100 | |||
| AK307502.1 | - | 12 - 13 | 1279 | 100 | ||
| BC126279.1 | - | 11 - 12 | 1335 | 8991995 | 100 | |
| BC126281.1 | - | 11 - 12 | 1335 | 8991997 | 100 | |
| FJ224324.1 | - | 11 - 12 | 1425 | 100 | ||
| X65873.1 | - | 11 - 12 | 1425 | 100 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001160.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
cytoplasm organization
axon guidance
cytoskeleton-dependent intracellular transport
positive regulation of synaptic transmission, GABAergic
stress granule disassembly
regulation of membrane potential
positive regulation of potassium ion transport
vesicle transport along microtubule
plus-end-directed vesicle transport along microtubule
positive regulation of establishment of protein localization to plasma membrane
cell-cell adhesion
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
cadherin binding involved in cell-cell adhesion
microtubule-based movement
cytoplasm organization
axon guidance
cytoskeleton-dependent intracellular transport
positive regulation of synaptic transmission, GABAergic
stress granule disassembly
regulation of membrane potential
positive regulation of potassium ion transport
vesicle transport along microtubule
plus-end-directed vesicle transport along microtubule
positive regulation of establishment of protein localization to plasma membrane
cell-cell adhesion
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR10014576.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR10014576.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR10014576.hg.1 | TC10001160.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000169790.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000169790.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000169790.hg.1 | ENST00000302418.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1000169790.hg.1 | NM_004521.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1000169790.hg.1 | TC1000010249.hg.1 | Clariom D TC |

KIF5B
Genomic Map