Assay Details
Gene Symbol: KIF5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_004521.2 | NP_004512.1 | 16 - 17 | 2381 | 146 | |
XM_017016224.1 | XP_016871713.1 | 16 - 17 | 1987 | 146 | ||
GenBank mRNA | AK289413.1 | - | 16 - 17 | 2327 | 146 | |
AK299542.1 | - | 752 | 146 | |||
BC126279.1 | - | 16 - 17 | 2134 | 8991995 | 146 | |
BC126281.1 | - | 16 - 17 | 2134 | 8991997 | 146 | |
FJ224324.1 | - | 16 - 17 | 2224 | 146 | ||
X65873.1 | - | 16 - 17 | 2224 | 146 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC10001160.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
microtubule-based movement
cytoplasm organization
axon guidance
cytoskeleton-dependent intracellular transport
positive regulation of synaptic transmission, GABAergic
stress granule disassembly
regulation of membrane potential
positive regulation of potassium ion transport
vesicle transport along microtubule
plus-end-directed vesicle transport along microtubule
positive regulation of establishment of protein localization to plasma membrane
cell-cell adhesion
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
cadherin binding involved in cell-cell adhesion
microtubule-based movement
cytoplasm organization
axon guidance
cytoskeleton-dependent intracellular transport
positive regulation of synaptic transmission, GABAergic
stress granule disassembly
regulation of membrane potential
positive regulation of potassium ion transport
vesicle transport along microtubule
plus-end-directed vesicle transport along microtubule
positive regulation of establishment of protein localization to plasma membrane
cell-cell adhesion
microtubule motor activity
protein binding
ATP binding
microtubule binding
ATP-dependent microtubule motor activity, plus-end-directed
cadherin binding involved in cell-cell adhesion
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10014570.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10014570.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10014570.hg.1 | TC10001160.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR10014571.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR10014571.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR10014571.hg.1 | TC10001160.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | PSR1000169784.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169784.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169784.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169784.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169784.hg.1 | TC1000010249.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | ENST00000493889.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1000010249.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169785.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169785.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169785.hg.1 | ENST00000302418.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1000169785.hg.1 | NM_004521.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1000169785.hg.1 | TC1000010249.hg.1 | Clariom D TC |
