Assay Details
| Gene Symbol: KIF5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004521.2 | NP_004512.1 | 21 - 22 | 2842 | 64 | |
| XM_017016224.1 | XP_016871713.1 | 21 - 22 | 2448 | 64 | ||
| GenBank mRNA | AK289413.1 | - | 21 - 22 | 2788 | 64 | |
| AK299542.1 | - | 1213 | 64 | |||
| BC126279.1 | - | 21 - 22 | 2595 | 8991995 | 64 | |
| BC126281.1 | - | 21 - 22 | 2595 | 8991997 | 64 | |
| FJ224324.1 | - | 21 - 22 | 2685 | 64 | ||
| X65873.1 | - | 21 - 22 | 2685 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC10001160.hg.1
Set Membership: |
|

KIF5B
Genomic Map