Assay Details
Gene Symbol: LSM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011528612.2 | XP_011526914.1 | 1 - 2 | 82 | 89 | |
GenBank mRNA | AK058202.1 | - | 1 - 2 | 87 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK058202.1, XM_011528612.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
