Assay Details
Gene Symbol: LSM14B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005260302.3 | XP_005260359.1 | 4 - 5 | 751 | 78 | |
XM_011528608.2 | XP_011526910.1 | 4 - 5 | 832 | 78 | ||
XM_011528613.2 | XP_011526915.1 | 4 - 5 | 473 | 78 | ||
XM_011528614.2 | XP_011526916.1 | 4 - 5 | 686 | 78 | ||
XM_017027690.1 | XP_016883179.1 | 2 - 3 | 287 | 78 | ||
GenBank mRNA | AK298748.1 | - | 3 - 4 | 348 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017027690.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279068.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000279069.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000361670.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000370915.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | ENST00000400318.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
Affymetrix Human Clariom D Assay | TC2000007991.hg.1 | NM_144703.2 | Refseq |
