Assay Details
| Gene Symbol: PIGU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080476.4 | NP_536724.1 | 7 - 8 | 834 | 93 | |
| XM_017027664.1 | XP_016883153.1 | 7 - 8 | 652 | 93 | ||
| GenBank mRNA | AB086842.1 | - | 7 - 8 | 640 | 93 | |
| AK075507.1 | - | 7 - 8 | 659 | 93 | ||
| AK301900.1 | - | 7 - 8 | 634 | 93 | ||
| AY339061.1 | - | 6 - 7 | 574 | 93 | ||
| AY358816.1 | - | 7 - 8 | 664 | 93 | ||
| AY422169.1 | - | 7 - 8 | 834 | 93 | ||
| BC030512.1 | - | 7 - 8 | 645 | 4942673 | 93 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC20000784.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR20010460.hg.1 | ENST00000217446.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010460.hg.1 | ENST00000374820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010460.hg.1 | ENST00000452740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010460.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010460.hg.1 | TC20000784.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010459.hg.1 | ENST00000217446.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010459.hg.1 | ENST00000374820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010459.hg.1 | ENST00000452740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010459.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010459.hg.1 | TC20000784.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010457.hg.1 | ENST00000217446.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010457.hg.1 | ENST00000374820.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010457.hg.1 | ENST00000452740.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010457.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010457.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR20010457.hg.1 | TC20000784.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | TC2000010001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171374.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171375.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171375.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171375.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171375.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171375.hg.1 | TC2000010001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171375.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171375.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171375.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171375.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171376.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171376.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171376.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2000171376.hg.1 | TC2000010001.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2000171376.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171376.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2000171376.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000217446.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000374820.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000438215.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000462389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | ENST00000480175.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC2000010001.hg.1 | NM_080476.4 | Refseq |

PIGU
Genomic Map