Assay Details
| Gene Symbol: ARPC5L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030978.2 | NP_112240.1 | 1 - 2 | 279 | 86 | |
| GenBank mRNA | AK056174.1 | - | 1 - 2 | 267 | 86 | |
| DA361505.1 | - | 1 - 2 | 279 | 86 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC09000646.hg
Additional Information:
This assay is in the 5'UTR region of the AK056174.1,NM_030978.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR09007408.hg.1 | TC09000646.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007408.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007408.hg.1 | ENST00000353214.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007408.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007409.hg.1 | TC09000646.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007409.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007409.hg.1 | ENST00000353214.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007409.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007411.hg.1 | TC09000646.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007411.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007411.hg.1 | ENST00000353214.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007411.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007411.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007412.hg.1 | TC09000646.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007412.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007412.hg.1 | ENST00000353214.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007412.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR09007412.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158299.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158299.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158299.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158299.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158299.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158299.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158299.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158299.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158299.hg.1 | TC0900008742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0900008742.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158294.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158294.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158294.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158294.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158294.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158294.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158294.hg.1 | TC0900008742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900158296.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158296.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158296.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158296.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158296.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158296.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158296.hg.1 | TC0900008742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900158295.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158295.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158295.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158295.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158295.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158295.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158295.hg.1 | TC0900008742.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0900158298.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158298.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158298.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158298.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158298.hg.1 | ENST00000259477.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158298.hg.1 | ENST00000353214.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158298.hg.1 | ENST00000465124.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0900158298.hg.1 | NM_030978.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0900158298.hg.1 | TC0900008742.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 160776 | - | - |

ARPC5L
Genomic Map