Assay Details
| Gene Symbol: SEC61A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013336.3 | NP_037468.1 | 5 - 6 | 541 | 75 | |
| GenBank mRNA | AF077032.1 | - | 5 - 6 | 447 | 75 | |
| AF084458.1 | - | 5 - 6 | 447 | 75 | ||
| AF346602.1 | - | 5 - 6 | 423 | 75 | ||
| AK027423.1 | - | 5 - 6 | 541 | 75 | ||
| AK074706.1 | - | 5 - 6 | 541 | 75 | ||
| AK299118.1 | - | 5 - 6 | 621 | 75 | ||
| AL832821.1 | - | 5 - 6 | 474 | 75 | ||
| BC002951.1 | - | 5 - 6 | 448 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03000670.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
SRP-dependent cotranslational protein targeting to membrane
posttranslational protein targeting to membrane
endoplasmic reticulum organization
cell growth
response to interferon-gamma
IRE1-mediated unfolded protein response
protein targeting to ER
protein binding
ribosome binding
SRP-dependent cotranslational protein targeting to membrane
posttranslational protein targeting to membrane
endoplasmic reticulum organization
cell growth
response to interferon-gamma
IRE1-mediated unfolded protein response
protein targeting to ER
protein binding
ribosome binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000498837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000498837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000498837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | ENST00000498837.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012066.hg.1 | TC03000670.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012066.hg.1 | ENST00000243253.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012066.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012066.hg.1 | ENST00000464451.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03012066.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300161953.hg.1 | TC0300008713.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000243253.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000424880.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000464451.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000481210.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000483956.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000491668.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000498837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000498837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000498837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | ENST00000498837.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300008713.hg.1 | NM_013336.3 | Refseq |

SEC61A1
Genomic Map