Assay Details
| Gene Symbol: DMTF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142326.1 | NP_001135798.1 | 7 - 8 | 886 | 84 | |
| NM_001142327.1 | NP_001135799.1 | 8 - 9 | 1003 | 84 | ||
| NM_021145.3 | NP_066968.3 | 10 - 11 | 1240 | 84 | ||
| XM_011516735.2 | XP_011515037.1 | 8 - 9 | 1925 | 84 | ||
| XM_011516737.1 | XP_011515039.1 | 9 - 10 | 1050 | 84 | ||
| XM_011516739.2 | XP_011515041.1 | 7 - 8 | 815 | 84 | ||
| XM_017012865.1 | XP_016868354.1 | 9 - 10 | 2019 | 84 | ||
| XM_017012866.1 | XP_016868355.1 | 7 - 8 | 1802 | 84 | ||
| XM_017012867.1 | XP_016868356.1 | 6 - 7 | 2470 | 84 | ||
| XM_017012868.1 | XP_016868357.1 | 9 - 10 | 1050 | 84 | ||
| XM_017012869.1 | XP_016868358.1 | 10 - 11 | 1079 | 84 | ||
| XM_017012870.1 | XP_016868359.1 | 7 - 8 | 818 | 84 | ||
| GenBank mRNA | AF084530.1 | - | 8 - 9 | 952 | 84 | |
| AK296211.1 | - | 7 - 8 | 779 | 84 | ||
| AK314622.1 | - | 8 - 9 | 939 | 84 | ||
| BC070064.1 | - | 10 - 11 | 1186 | 4373991 | 84 | |
More Information
Set Membership: |
|

DMTF1
Genomic Map