Assay Details
Gene Symbol: NRXN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_073547.1 | 9 - 10 | 3242 | 79 | ||
XM_017021793.1 | XP_016877282.1 | 11 - 12 | 3284 | 79 | ||
XM_017021796.1 | XP_016877285.1 | 10 - 11 | 3260 | 79 | ||
XM_017021798.1 | XP_016877287.1 | 9 - 10 | 3248 | 79 | ||
XM_017021801.1 | XP_016877290.1 | 11 - 12 | 3284 | 79 | ||
XM_017021804.1 | XP_016877293.1 | 11 - 12 | 3284 | 79 | ||
XM_017021806.1 | XP_016877295.1 | 10 - 11 | 3260 | 79 | ||
GenBank mRNA | AJ316284.2 | - | 9 - 10 | 3030 | 79 | |
AK310235.1 | - | 6 - 7 | 1587 | 79 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
