Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023628.hg.1 |
TC06001928.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023628.hg.1 |
ENST00000369523.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023628.hg.1 |
ENST00000369536.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06023628.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000493269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000493269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000493269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000493269.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000497828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000497828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000497828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
ENST00000497828.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186194.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186194.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186194.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600186194.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600186194.hg.1 |
TC0600012512.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000369536.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000451155.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000493269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000493269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000493269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000493269.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000497828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000497828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000497828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
ENST00000497828.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600012512.hg.1 |
NM_020320.4 |
Refseq |