Assay Details
| Gene Symbol: CNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160226.1 | NP_001153698.1 | 2 - 3 | 392 | 87 | |
| NM_001160258.1 | NP_001153730.1 | 3 - 4 | 430 | 87 | ||
| XM_017010238.1 | XP_016865727.1 | 3 - 4 | 3560 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001160226.1, NM_001160258.1, XM_017010238.1 transcripts.
Set Membership: |
|

CNR1
Genomic Map