Assay Details
| Gene Symbol: FAAH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001441.2 | NP_001432.2 | 12 - 13 | 1440 | 71 | |
| GenBank mRNA | BC093632.1 | - | 12 - 13 | 1382 | 7939477 | 71 |
| BC110404.1 | - | 12 - 13 | 1370 | 5728192 | 71 | |
| BC111941.1 | - | 12 - 13 | 1382 | 8327409 | 71 | |
| U82535.1 | - | 12 - 13 | 1393 | 71 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000585.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01010010.hg.1 | TC01000585.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010010.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010010.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493636.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158398.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158398.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158398.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158398.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158398.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158398.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158398.hg.1 | TC0100008158.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000243167.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000468718.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000484697.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000489366.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493636.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | ENST00000493735.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008158.hg.1 | NM_001441.2 | Refseq |

FAAH
Genomic Map