Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011542805.1 | XP_011541107.1 | 1 - 2 | 205 | 74 | |
| XM_011542806.2 | XP_011541108.1 | 1 - 2 | 201 | 74 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011542805.1, XM_011542806.2 transcripts.
Set Membership: |
|

IL18
Genomic Map