Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243211.1 | NP_001230140.1 | 2 - 3 | 296 | 143 | |
| XM_011542805.1 | XP_011541107.1 | 2 - 3 | 304 | 143 | ||
| GenBank mRNA | AY266351.1 | - | 2 - 3 | 253 | 143 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11002293.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
angiogenesis
inflammatory response
immune response
cell-cell signaling
positive regulation of gene expression
natural killer cell activation
regulation of cell adhesion
sleep
lipopolysaccharide-mediated signaling pathway
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-17 production
positive regulation of natural killer cell proliferation
positive regulation of tissue remodeling
chemokine biosynthetic process
T-helper 1 type immune response
type 2 immune response
interleukin-2 biosynthetic process
interferon-gamma biosynthetic process
positive regulation of activated T cell proliferation
interleukin-13 biosynthetic process
granulocyte macrophage colony-stimulating factor biosynthetic process
positive regulation of NF-kappaB import into nucleus
positive regulation of tyrosine phosphorylation of Stat3 protein
negative regulation of myoblast differentiation
positive regulation of inflammatory response
positive regulation of NK T cell proliferation
cellular response to organic cyclic compound
cytokine activity
protein binding
MAPK cascade
angiogenesis
inflammatory response
immune response
cell-cell signaling
positive regulation of gene expression
natural killer cell activation
regulation of cell adhesion
sleep
lipopolysaccharide-mediated signaling pathway
positive regulation of granulocyte macrophage colony-stimulating factor production
positive regulation of interferon-gamma production
positive regulation of interleukin-17 production
positive regulation of natural killer cell proliferation
positive regulation of tissue remodeling
chemokine biosynthetic process
T-helper 1 type immune response
type 2 immune response
interleukin-2 biosynthetic process
interferon-gamma biosynthetic process
positive regulation of activated T cell proliferation
interleukin-13 biosynthetic process
granulocyte macrophage colony-stimulating factor biosynthetic process
positive regulation of NF-kappaB import into nucleus
positive regulation of tyrosine phosphorylation of Stat3 protein
negative regulation of myoblast differentiation
positive regulation of inflammatory response
positive regulation of NK T cell proliferation
cellular response to organic cyclic compound
cytokine activity
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000534225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000534225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | ENST00000534225.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027480.hg.1 | ENST00000280357.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027480.hg.1 | ENST00000524595.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027480.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027480.hg.1 | ENST00000533858.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027480.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027480.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11027480.hg.1 | TC11002293.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000534225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000534225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | ENST00000534225.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100012318.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000280357.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000524595.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000525547.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000528832.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | ENST00000533858.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | NM_001243211.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | NM_001562.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100194678.hg.1 | TC1100012318.hg.1 | Clariom D TC |

IL18
Genomic Map