Assay Details
Gene Symbol: PPP2CB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001009552.1 | NP_001009552.1 | 3 - 4 | 904 | 129 | |
GenBank mRNA | AF444006.1 | - | 3 - 4 | 490 | 129 | |
BC012022.1 | - | 3 - 4 | 515 | 4454972 | 129 | |
BT019526.1 | - | 3 - 4 | 489 | 129 | ||
CR541747.1 | - | 3 - 4 | 489 | 129 | ||
J03805.1 | - | 3 - 4 | 487 | 129 | ||
X12656.1 | - | 3 - 4 | 510 | 129 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC08001111.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
protein dephosphorylation
apoptotic mitochondrial changes
regulation of gene expression
response to endoplasmic reticulum stress
response to hydrogen peroxide
proteasome-mediated ubiquitin-dependent protein catabolic process
response to antibiotic
protein serine/threonine phosphatase activity
protein binding
metal ion binding
protein dephosphorylation
apoptotic mitochondrial changes
regulation of gene expression
response to endoplasmic reticulum stress
response to hydrogen peroxide
proteasome-mediated ubiquitin-dependent protein catabolic process
response to antibiotic
protein serine/threonine phosphatase activity
protein binding
metal ion binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR08014354.hg.1 | TC08001111.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08014354.hg.1 | ENST00000221138.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014354.hg.1 | ENST00000406655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014354.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000520500.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000522113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000522113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000522113.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000523023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000523023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000523023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000523023.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR08014356.hg.1 | TC08001111.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR08014356.hg.1 | ENST00000221138.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014356.hg.1 | ENST00000406655.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR08014356.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168359.hg.1 | TC0800010043.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0800168357.hg.1 | TC0800010043.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000221138.8 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518243.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518532.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000518564.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520056.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520334.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000520500.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000522113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000522113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000522113.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000523023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000523023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000523023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000523023.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | ENST00000523804.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0800010043.hg.1 | NM_001009552.1 | Refseq |
