Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018029.hg.1 |
TC0X001347.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018029.hg.1 |
ENST00000371130.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018029.hg.1 |
ENST00000422452.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018029.hg.1 |
NM_001163278.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018029.hg.1 |
NM_001163279.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR0X018029.hg.1 |
NM_014253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
ENST00000371130.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
ENST00000422452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
NM_001163278.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
NM_001163279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
NM_014253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
ENST00000371130.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
ENST00000422452.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
NM_001163278.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
NM_001163279.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
NM_014253.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0X00172952.hg.1 |
TC0X00010734.hg.1 |
Clariom D TC |