Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008495.hg.1 |
ENST00000238961.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008495.hg.1 |
ENST00000370269.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008495.hg.1 |
NM_001136123.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008495.hg.1 |
NM_018121.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008495.hg.1 |
TC10000726.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1000159093.hg.1 |
ENST00000238961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159093.hg.1 |
ENST00000370269.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159093.hg.1 |
NM_001136123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159093.hg.1 |
NM_018121.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159093.hg.1 |
ENST00000238961.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159093.hg.1 |
ENST00000370269.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000159093.hg.1 |
NM_001136123.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159093.hg.1 |
NM_018121.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000159093.hg.1 |
TC1000008661.hg.1 |
Clariom D TC |