Assay Details
| Gene Symbol: SPATA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138796.3 | NP_620151.1 | 1 - 2 | 120 | 99 | |
| XM_005273052.1 | XP_005273109.1 | 1 - 2 | 102 | 99 | ||
| XM_006711165.3 | XP_006711228.1 | 1 - 2 | 111 | 99 | ||
| XM_011509194.2 | XP_011507496.1 | 1 - 2 | 102 | 99 | ||
| GenBank mRNA | AK098591.1 | - | 1 - 2 | 100 | 99 | |
| AK310291.1 | - | 1 - 2 | 104 | 99 | ||
| AY963797.1 | - | 1 - 2 | 123 | 99 | ||
| BC014608.2 | - | 1 - 2 | 91 | 4824429 | 99 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001799.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028115.hg.1 | TC01001799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028115.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028115.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028114.hg.1 | TC01001799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028114.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028114.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185481.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185481.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185481.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185481.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185481.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185481.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185481.hg.1 | TC0100011611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100185482.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185482.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185482.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185482.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185482.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185482.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185482.hg.1 | TC0100011611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100185480.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185480.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185480.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185480.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185480.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185480.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185480.hg.1 | TC0100011611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185479.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185479.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185479.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185479.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185479.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185479.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185479.hg.1 | TC0100011611.hg.1 | Clariom D TC |

SPATA17
Genomic Map