Assay Details
| Gene Symbol: SPATA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_138796.3 | NP_620151.1 | 9 - 10 | 1062 | 135 | |
| GenBank mRNA | AK098591.1 | - | 9 - 10 | 1042 | 135 | |
| AY963797.1 | - | 9 - 10 | 1065 | 135 | ||
| BC014608.2 | - | 9 - 10 | 1033 | 4824429 | 135 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001799.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01028131.hg.1 | TC01001799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028131.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028131.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028131.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028133.hg.1 | TC01001799.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028133.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028133.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01028133.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185495.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185495.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185495.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185495.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185495.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185495.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185495.hg.1 | TC0100011611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100185494.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185494.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185494.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185494.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185494.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185494.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185494.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185494.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185494.hg.1 | TC0100011611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100185497.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185497.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185497.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185497.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185497.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185497.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185497.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100185497.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100185497.hg.1 | TC0100011611.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000366933.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000470448.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000471021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000491809.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | ENST00000492747.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100011611.hg.1 | NM_138796.3 | Refseq |

SPATA17
Genomic Map