Assay Details
| Gene Symbol: ZNF273 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC007078.1 | - | 1 - 2 | 113 | 70 | |
| BC019234.1 | - | 1 - 2 | 117 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC019234.1 transcripts.
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000395375.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000476120.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | ENST00000489672.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NM_021148.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0700013375.hg.1 | NR_003099.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 227710 | - | - |

ZNF273
Genomic Map