Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027070.hg.1 |
TC06002146.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027070.hg.1 |
ENST00000355845.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027070.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027070.hg.1 |
ENST00000367768.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR06027070.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000463140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000463140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
ENST00000463140.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191948.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191948.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191948.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600191948.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600191948.hg.1 |
TC0600013280.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000359015.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000463140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000463140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
ENST00000463140.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600013280.hg.1 |
NM_005923.3 |
Refseq |