Assay Details
Gene Symbol: SAP30L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024632.5 | NP_078908.1 | 2 - 3 | 972 | 53 | |
GenBank mRNA | AK021588.1 | - | 2 - 3 | 972 | 53 | |
AY341060.1 | - | 2 - 3 | 915 | 53 | ||
AY846876.1 | - | 2 - 3 | 324 | 53 | ||
BC009829.2 | - | 2 - 3 | 906 | 3937610 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0900008340.hg.1 | ENST00000417622.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000519683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000519683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000519683.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0900008340.hg.1 | ENST00000417622.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000297109.10 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000426761.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000440364.6 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000517926.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000519683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000519683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000519683.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520159.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000520731.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523029.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | ENST00000523198.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131062.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_001131063.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NM_024632.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0500009161.hg.1 | NR_024084.1 | Refseq |
