Assay Details
| Gene Symbol: NAT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020378.3 | NP_065111.1 | 1 - 2 | 67 | 125 | |
| GenBank mRNA | BM785191.1 | - | 1 - 2 | 69 | 125 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC19000899.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_020378.3 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR19012476.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012476.hg.1 | TC19000899.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000588985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000588985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000588985.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012477.hg.1 | ENST00000205194.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012477.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012477.hg.1 | TC19000899.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012478.hg.1 | ENST00000205194.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012478.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR19012478.hg.1 | TC19000899.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000588985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000588985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000588985.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1900008911.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | TC1900008911.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169853.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900169849.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169849.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169849.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900169849.hg.1 | TC1900008911.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900169849.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169849.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169849.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | NM_020378.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | TC1900008911.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | ENST00000205194.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | ENST00000587400.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | ENST00000591590.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | ENST00000592719.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1900169852.hg.1 | NM_020378.3 | Refseq |

NAT14
Genomic Map