Assay Details
| Gene Symbol: MRPL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014763.3 | NP_055578.2 | 4 - 5 | 501 | 87 | |
| GenBank mRNA | AK291250.1 | - | 4 - 5 | 487 | 87 | |
| AK295578.1 | - | 4 - 5 | 473 | 87 | ||
| BC030144.2 | - | 4 - 5 | 486 | 4763405 | 87 | |
| BI551514.1 | - | 4 - 5 | 507 | 87 | ||
| BQ432684.1 | - | 4 - 5 | 504 | 87 | ||
| D14660.1 | - | 4 - 5 | 474 | 87 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02000476.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02007415.hg.1 | TC02000476.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007415.hg.1 | ENST00000393909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007415.hg.1 | ENST00000409374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007415.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007416.hg.1 | TC02000476.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007416.hg.1 | ENST00000393909.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007416.hg.1 | ENST00000409374.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007416.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02007416.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | ENST00000604845.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000358788.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000453233.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000493686.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | ENST00000604845.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200008128.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156434.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156434.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156434.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156434.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156434.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156434.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156434.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156434.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156434.hg.1 | TC0200008128.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | ENST00000393909.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | ENST00000409374.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | ENST00000476622.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | ENST00000492255.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | NM_014763.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200156435.hg.1 | TC0200008128.hg.1 | Clariom D TC |

MRPL19
Genomic Map