Assay Details
Gene Symbol: LRRC71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005244926.3 | XP_005244983.1 | 12 - 13 | 1521 | 60 | |
XM_006711185.3 | XP_006711248.1 | 13 - 14 | 1311 | 60 | ||
XM_006711186.3 | XP_006711249.1 | 12 - 13 | 1521 | 60 | ||
XM_011509239.2 | XP_011507541.1 | 8 - 9 | 993 | 60 | ||
XM_011509240.2 | XP_011507542.1 | 8 - 9 | 975 | 60 | ||
XM_017000460.1 | XP_016855949.1 | 12 - 13 | 1829 | 60 | ||
XM_017000461.1 | XP_016855950.1 | 13 - 14 | 1367 | 60 | ||
XM_017000462.1 | XP_016855951.1 | 12 - 13 | 1270 | 60 | ||
GenBank mRNA | AK057446.1 | - | 13 - 14 | 1326 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000472465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000472465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000472465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000472465.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000337428.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000472465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000472465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000472465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000472465.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000476550.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | ENST00000490146.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100010198.hg.1 | NM_144702.2 | Refseq |
