Assay Details
| Gene Symbol: SREK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270492.1 | NP_001257421.1 | 11 - 12 | 114 | ||
| NM_001323529.1 | NP_001310458.1 | 10 - 11 | 114 | |||
| NM_001323535.1 | NP_001310464.1 | 10 - 11 | 114 | |||
| GenBank mRNA | AK308287.1 | - | 11 - 12 | 1762 | 113 | |
More Information
Set Membership: |
|

SREK1
Genomic Map