Assay Details
Gene Symbol: CALML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | M36707.1 | - | 1 | 2681 | 76 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | NM_005185.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | NM_005185.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | NM_005185.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1000006624.hg.1 | NM_005185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | ENST00000315238.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | NM_005185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | NM_005185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | NM_005185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000006624.hg.1 | NM_005185.3 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000542093.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000543008.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | ENST00000545372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120496.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1000009667.hg.1 | NR_120497.1 | Refseq |
