Assay Details
Gene Symbol: FAM186B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_032130.2 | NP_115506.1 | 4 - 5 | 2342 | 98 | |
NR_027450.1 | 4 - 5 | 2499 | 98 | |||
XM_006719625.2 | XP_006719688.1 | 4 - 5 | 2331 | 98 | ||
XM_006719626.2 | XP_006719689.1 | 4 - 5 | 2331 | 98 | ||
XM_006719627.3 | XP_006719690.1 | 4 - 5 | 2331 | 98 | ||
XM_011538796.2 | XP_011537098.1 | 4 - 5 | 2331 | 98 | ||
XM_011538797.2 | XP_011537099.1 | 4 - 5 | 2331 | 98 | ||
XM_017020008.1 | XP_016875497.1 | 4 - 5 | 2582 | 98 | ||
GenBank mRNA | AK093242.1 | - | 4 - 5 | 2331 | 98 | |
AK302702.1 | - | 4 - 5 | 2499 | 98 | ||
AL136748.1 | - | 4 - 5 | 2263 | 98 | ||
AL713673.1 | - | 1 - 2 | 1008 | 98 | ||
BC035621.1 | - | 4 - 5 | 2300 | 5171752 | 98 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001480.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000527253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000527253.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019431.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019431.hg.1 | ENST00000544141.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR12019431.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019431.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR12019431.hg.1 | TC12001480.hg.1 | HTA TC |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000508736.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000527253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000527253.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000547764.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200012625.hg.1 | ENST00000552301.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000548841.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | ENST00000551047.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | TC1200010635.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | ENST00000257894.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | ENST00000532262.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | ENST00000533372.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | NM_032130.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | NR_027450.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR1200180897.hg.1 | TC1200010635.hg.1 | Clariom D TC |
