Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
ENST00000335753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
ENST00000369755.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
ENST00000474260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
NM_001304743.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
NM_014720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
ENST00000335753.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
ENST00000369755.3 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
ENST00000474260.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
NM_001304743.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
NM_014720.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000160182.hg.1 |
TC1000008795.hg.1 |
Clariom D TC |