Assay Details
| Gene Symbol: RIPK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003804.4 | NP_003795.2 | 4 - 5 | 695 | 121 | |
| XM_005249457.3 | XP_005249514.2 | 4 - 5 | 618 | 121 | ||
| XM_006715237.3 | XP_006715300.2 | 4 - 5 | 618 | 121 | ||
| XM_017011405.1 | XP_016866894.1 | 4 - 5 | 616 | 121 | ||
| GenBank mRNA | AK314176.1 | - | 4 - 5 | 695 | 121 | |
| BC126254.1 | - | 3 - 4 | 489 | 8991970 | 121 | |
| BC126256.1 | - | 3 - 4 | 489 | 8991972 | 121 | |
| U50062.1 | - | 3 - 4 | 463 | 121 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06000023.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of protein phosphorylation
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
I-kappaB kinase/NF-kappaB signaling
activation of JUN kinase activity
regulation of tumor necrosis factor-mediated signaling pathway
regulation of necrotic cell death
positive regulation of necrotic cell death
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
tumor necrosis factor-mediated signaling pathway
response to tumor necrosis factor
TRIF-dependent toll-like receptor signaling pathway
peptidyl-serine autophosphorylation
positive regulation of apoptotic process
positive regulation of programmed cell death
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
cellular protein catabolic process
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
protein autophosphorylation
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of necroptotic process
T cell apoptotic process
necroptotic process
regulation of ATP:ADP antiporter activity
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
death-inducing signaling complex assembly
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
ripoptosome assembly
necroptotic signaling pathway
ripoptosome assembly involved in necroptotic process
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of hydrogen peroxide-induced cell death
amyloid fibril formation
positive regulation of reactive oxygen species metabolic process
negative regulation of extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
death receptor binding
protein binding
ATP binding
ubiquitin protein ligase binding
protein complex binding
identical protein binding
death domain binding
positive regulation of protein phosphorylation
apoptotic process
activation of cysteine-type endopeptidase activity involved in apoptotic process
I-kappaB kinase/NF-kappaB signaling
activation of JUN kinase activity
regulation of tumor necrosis factor-mediated signaling pathway
regulation of necrotic cell death
positive regulation of necrotic cell death
positive regulation of interleukin-8 production
positive regulation of tumor necrosis factor production
tumor necrosis factor-mediated signaling pathway
response to tumor necrosis factor
TRIF-dependent toll-like receptor signaling pathway
peptidyl-serine autophosphorylation
positive regulation of apoptotic process
positive regulation of programmed cell death
positive regulation of I-kappaB kinase/NF-kappaB signaling
negative regulation of I-kappaB kinase/NF-kappaB signaling
cellular protein catabolic process
positive regulation of macrophage differentiation
positive regulation of transcription from RNA polymerase II promoter
positive regulation of JNK cascade
protein autophosphorylation
positive regulation of NF-kappaB transcription factor activity
protein homooligomerization
protein heterooligomerization
positive regulation of necroptotic process
T cell apoptotic process
necroptotic process
regulation of ATP:ADP antiporter activity
cellular response to tumor necrosis factor
cellular response to growth factor stimulus
death-inducing signaling complex assembly
apoptotic signaling pathway
extrinsic apoptotic signaling pathway
activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
ripoptosome assembly
necroptotic signaling pathway
ripoptosome assembly involved in necroptotic process
regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
positive regulation of hydrogen peroxide-induced cell death
amyloid fibril formation
positive regulation of reactive oxygen species metabolic process
negative regulation of extrinsic apoptotic signaling pathway
positive regulation of extrinsic apoptotic signaling pathway
negative regulation of extrinsic apoptotic signaling pathway in absence of ligand
protein kinase activity
protein serine/threonine kinase activity
death receptor binding
protein binding
ATP binding
ubiquitin protein ligase binding
protein complex binding
identical protein binding
death domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000172.hg.1 | TC06000023.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000172.hg.1 | ENST00000259808.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000172.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000172.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000172.hg.1 | ENST00000541791.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR06000172.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145561.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145561.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145561.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145561.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145561.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145561.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145561.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145561.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145561.hg.1 | TC0600006653.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0600145563.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145563.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145563.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145563.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145563.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145563.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145563.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0600145563.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0600145563.hg.1 | TC0600006653.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000259808.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000380409.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | ENST00000479389.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0600006653.hg.1 | NM_003804.5 | Refseq |

RIPK1
Genomic Map