Assay Details
| Gene Symbol: ADAM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003183.5 | NP_003174.3 | 5 - 6 | 836 | 72 | |
| XM_011510375.2 | XP_011508677.1 | 5 - 6 | 855 | 72 | ||
| XM_017004785.1 | XP_016860274.1 | 5 - 6 | 852 | 72 | ||
| GenBank mRNA | BC136783.1 | - | 5 - 6 | 698 | 9020773 | 72 |
| BC146658.1 | - | 5 - 6 | 808 | 72 | ||
| GQ402545.1 | - | 5 - 6 | 804 | 72 | ||
| U69611.1 | - | 5 - 6 | 735 | 72 | ||
| U69612.1 | - | 5 - 6 | 743 | 72 | ||
| U86755.1 | - | 5 - 6 | 784 | 72 | ||
| U92649.1 | - | 5 - 6 | 785 | 72 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001558.hg.1
Additional Information:
This assay is in the 5'UTR region of the XM_017004785.1 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
response to hypoxia
positive regulation of protein phosphorylation
neutrophil mediated immunity
germinal center formation
positive regulation of leukocyte chemotaxis
proteolysis
membrane protein ectodomain proteolysis
cell adhesion
epidermal growth factor receptor signaling pathway
Notch signaling pathway
Notch receptor processing
positive regulation of cell proliferation
positive regulation of T cell chemotaxis
B cell differentiation
positive regulation of cell growth
positive regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
membrane protein intracellular domain proteolysis
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
response to lipopolysaccharide
positive regulation of chemokine production
regulation of mast cell apoptotic process
T cell differentiation in thymus
tumor necrosis factor-mediated signaling pathway
cell adhesion mediated by integrin
wound healing, spreading of epidermal cells
epidermal growth factor-activated receptor transactivation by G-protein coupled receptor signaling pathway
response to drug
positive regulation of epidermal growth factor-activated receptor activity
spleen development
cell motility
defense response to Gram-positive bacterium
PMA-inducible membrane protein ectodomain proteolysis
positive regulation of cellular component movement
response to high density lipoprotein particle
metalloendopeptidase activity
Notch binding
interleukin-6 receptor binding
integrin binding
protein binding
metallopeptidase activity
zinc ion binding
SH3 domain binding
PDZ domain binding
response to hypoxia
positive regulation of protein phosphorylation
neutrophil mediated immunity
germinal center formation
positive regulation of leukocyte chemotaxis
proteolysis
membrane protein ectodomain proteolysis
cell adhesion
epidermal growth factor receptor signaling pathway
Notch signaling pathway
Notch receptor processing
positive regulation of cell proliferation
positive regulation of T cell chemotaxis
B cell differentiation
positive regulation of cell growth
positive regulation of cell migration
positive regulation of transforming growth factor beta receptor signaling pathway
negative regulation of transforming growth factor beta receptor signaling pathway
membrane protein intracellular domain proteolysis
positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
response to lipopolysaccharide
positive regulation of chemokine production
regulation of mast cell apoptotic process
T cell differentiation in thymus
tumor necrosis factor-mediated signaling pathway
cell adhesion mediated by integrin
wound healing, spreading of epidermal cells
epidermal growth factor-activated receptor transactivation by G-protein coupled receptor signaling pathway
response to drug
positive regulation of epidermal growth factor-activated receptor activity
spleen development
cell motility
defense response to Gram-positive bacterium
PMA-inducible membrane protein ectodomain proteolysis
positive regulation of cellular component movement
response to high density lipoprotein particle
metalloendopeptidase activity
Notch binding
interleukin-6 receptor binding
integrin binding
protein binding
metallopeptidase activity
zinc ion binding
SH3 domain binding
PDZ domain binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02025251.hg.1 | TC02001558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025251.hg.1 | ENST00000310823.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025251.hg.1 | ENST00000497134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025251.hg.1 | ENST00000538558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025251.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025250.hg.1 | TC02001558.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025250.hg.1 | ENST00000310823.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025250.hg.1 | ENST00000497134.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025250.hg.1 | ENST00000538558.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02025250.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000478059.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200011688.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184961.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184961.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184961.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184961.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184961.hg.1 | ENST00000310823.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184961.hg.1 | ENST00000497134.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184961.hg.1 | ENST00000618923.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200184961.hg.1 | NM_003183.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200184961.hg.1 | TC0200011688.hg.1 | Clariom D TC |

ADAM17
Genomic Map