Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017758.hg.1 |
ENST00000265981.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017758.hg.1 |
ENST00000528665.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017758.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR11017758.hg.1 |
TC11001414.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000534281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000534281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000534281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
ENST00000534281.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1100176469.hg.1 |
TC1100010089.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000265981.6 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000528665.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000530156.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000533412.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000534281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000534281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000534281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
ENST00000534281.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1100010089.hg.1 |
NM_016422.3 |
Refseq |