Assay Details
| Gene Symbol: HOOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015888.4 | NP_056972.1 | 13 - 14 | 1587 | 83 | |
| XM_006710676.1 | XP_006710739.1 | 13 - 14 | 1527 | 83 | ||
| XM_011541562.2 | XP_011539864.1 | 13 - 14 | 1557 | 83 | ||
| XM_011541563.1 | XP_011539865.1 | 13 - 14 | 1527 | 83 | ||
| GenBank mRNA | AF044923.1 | - | 13 - 14 | 1529 | 83 | |
| AK097735.1 | - | 13 - 14 | 1448 | 83 | ||
| AK098561.1 | - | 9 - 10 | 986 | 83 | ||
| AK292314.1 | - | 14 - 15 | 1432 | 83 | ||
| AK301768.1 | - | 13 - 14 | 1355 | 83 | ||
| BC011621.2 | - | 13 - 14 | 1587 | 3959931 | 83 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000700.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01011149.hg.1 | TC01000700.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011149.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011149.hg.1 | ENST00000395561.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011149.hg.1 | ENST00000465876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011149.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160462.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160462.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160462.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160462.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160462.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160462.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160462.hg.1 | TC0100008503.hg.1 | Clariom D TC |

HOOK1
Genomic Map