Assay Details
Gene Symbol: HOOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015888.4 | NP_056972.1 | 14 - 15 | 1645 | 142 | |
XM_006710676.1 | XP_006710739.1 | 14 - 15 | 1585 | 142 | ||
XM_011541562.2 | XP_011539864.1 | 14 - 15 | 1615 | 142 | ||
XM_011541563.1 | XP_011539865.1 | 14 - 15 | 1585 | 142 | ||
GenBank mRNA | AF044923.1 | - | 14 - 15 | 1587 | 142 | |
AK097735.1 | - | 14 - 15 | 1506 | 142 | ||
AK098561.1 | - | 10 - 11 | 1044 | 142 | ||
AK292314.1 | - | 15 - 16 | 1490 | 142 | ||
AK301768.1 | - | 14 - 15 | 1413 | 142 | ||
BC011621.2 | - | 14 - 15 | 1645 | 3959931 | 142 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000700.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01011150.hg.1 | TC01000700.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01011150.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011150.hg.1 | ENST00000395561.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011150.hg.1 | ENST00000465876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011150.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160463.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160463.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160463.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160463.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160463.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160463.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160463.hg.1 | TC0100008503.hg.1 | Clariom D TC |
