Assay Details
| Gene Symbol: HOOK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015888.4 | NP_056972.1 | 21 - 22 | 2276 | 121 | |
| XM_011541562.2 | XP_011539864.1 | 21 - 22 | 2246 | 121 | ||
| GenBank mRNA | AF044923.1 | - | 21 - 22 | 2218 | 121 | |
| AK097735.1 | - | 19 - 20 | 2019 | 121 | ||
| AK098561.1 | - | 15 - 16 | 1557 | 121 | ||
| AK292314.1 | - | 22 - 23 | 2121 | 121 | ||
| AK301768.1 | - | 21 - 22 | 2044 | 121 | ||
| BC011621.2 | - | 21 - 22 | 2276 | 3959931 | 121 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000700.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01011167.hg.1 | TC01000700.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011167.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011167.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011162.hg.1 | TC01000700.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011162.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011162.hg.1 | ENST00000395561.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011162.hg.1 | ENST00000465876.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01011162.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | ENST00000465876.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | ENST00000466803.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | ENST00000491135.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160476.hg.1 | TC0100008503.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100160481.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160481.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160481.hg.1 | ENST00000371208.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100160481.hg.1 | NM_015888.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100160481.hg.1 | TC0100008503.hg.1 | Clariom D TC |

HOOK1
Genomic Map