Assay Details
Gene Symbol: HOOK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015888.4 | NP_056972.1 | 6 - 7 | 724 | 82 | |
XM_006710676.1 | XP_006710739.1 | 6 - 7 | 664 | 82 | ||
XM_011541562.2 | XP_011539864.1 | 6 - 7 | 694 | 82 | ||
XM_011541563.1 | XP_011539865.1 | 6 - 7 | 664 | 82 | ||
XM_017001424.1 | XP_016856913.1 | 6 - 7 | 664 | 82 | ||
GenBank mRNA | AF044923.1 | - | 6 - 7 | 666 | 82 | |
AK097735.1 | - | 6 - 7 | 585 | 82 | ||
AK098561.1 | - | 2 - 3 | 122 | 82 | ||
AK292314.1 | - | 7 - 8 | 569 | 82 | ||
AK301768.1 | - | 6 - 7 | 492 | 82 | ||
BC011621.2 | - | 6 - 7 | 724 | 3959931 | 82 | |
BC107857.1 | - | 6 - 7 | 638 | 82 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000700.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR01011142.hg.1 | TC01000700.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01011142.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011142.hg.1 | ENST00000395561.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011142.hg.1 | ENST00000465876.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01011142.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160455.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160455.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160455.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160455.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160455.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100160455.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100160455.hg.1 | TC0100008503.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000371208.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000455990.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000465876.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000466803.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000474695.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | ENST00000491135.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100008503.hg.1 | NM_015888.4 | Refseq |
